missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eisco™ Set of Masses with Hanger
For classrooms and labs in demonstrating force and balance.
$122.32
Spécifications
| Product Type | Mass and Hanger Set |
|---|---|
| Material | Stainless Steel |
| For Use With (Application) | Balance testing; Weight determination; Classroom tool for demonstrations on force and balance |
| Numéro de catalogue | Numéro du manufacturier. | Weight (Metric) | Prix | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Weight (Metric) | Prix | Quantity & Availability | |||||
|
S24419
|
Eisco™
PH0258MSS |
250 g |
chaque for $122.32
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
Description
- Weight set made from stainless-steel
- Hangers with hook
- Masses removable and replaceble on hanger
Spécifications
| Mass and Hanger Set | |
| Stainless Steel | |
| Balance testing; Weight determination; Classroom tool for demonstrations on force and balance |