missing translation for 'onlineSavingsMsg'
Learn More

Sesamol 98.0+%, TCI America™

Catalog No. S041825G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S041825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S041825G Supplier TCI America Supplier No. S041825G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 533-31-3
Molecular Formula C7H6O3
Molecular Weight (g/mol) 138.122
MDL Number MFCD00005827
InChI Key LUSZGTFNYDARNI-UHFFFAOYSA-N
Synonym sesamol, 3,4-methylenedioxy phenol, 5-hydroxy-1,3-benzodioxole, 3,4-methylenedioxyphenol, 2h-1,3-benzodioxol-5-ol, 3,4-methylendioxyphenol, methylene ether of oxyhydroquinone, phenol, 3,4-methylenedioxy, 5-benzodioxolol, unii-94iea0nv89
PubChem CID 68289
ChEBI CHEBI:9126
IUPAC Name 1,3-benzodioxol-5-ol
SMILES C1OC2=C(O1)C=C(C=C2)O

Specifications

Melting Point 64°C
Boiling Point 113°C
Quantity 25 g
Formula Weight 138.12
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Sesamol
EINECSNumber (5)-6772
RTECSNumber SM0890000
TSCA Yes