missing translation for 'onlineSavingsMsg'
Learn More

Sesame oil

Catalog No. AC241002500
Change view
Click to view available options
Quantity:
250 mL
1 L
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC241002500 250 mL Glass bottle
AC241000010 1 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC241002500 Supplier Thermo Scientific Chemicals Supplier No. 241002500
Only null left

CAS: 8008-74-0

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 8008-74-0
MDL Number MFCD00132243

Specifications

Acidity 0.6mg KOH/g max.
Melting Point -5.0°C
Color Yellow
Density 0.9190g/mL
Flash Point 255°C
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4700 to 1.4760
Quantity 250 mL
Specific Gravity 0.919
Merck Index 14, 8469
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in chloroform, ether, petroleum ethers
Physical Form Viscous Liquid
Chemical Name or Material Sesame oil
Show More Show Less
EINECSNumber 232-370-6
RTECSNumber VU3940000
TSCA TSCA

RUO – Research Use Only