Learn More
METTLER TOLEDO™ Service for Excellence XSR Balances
Numéro de catalogue. 01670024
Change view
Click to view available options
Includes:
EQPac Regulated Balances with USP
IPac Balances with Accredited ACC
StarterPac Installation with ACC
For Use With (Equipment):
Excellence XSR Analytical Balances
Excellence XSR Precision Balances
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Includes For Use With (Equipment)
01670024 EQPac Regulated Balances with USP Excellence XSR Precision Balances
01670008 IPac Balances with Accredited ACC Excellence XSR Precision Balances
01670020 StarterPac Installation with ACC Excellence XSR Analytical Balances
01670040 IPac Balances with Accredited ACC Excellence XSR Analytical Balances
01670041 EQPac Regulated Balances with USP Excellence XSR Analytical Balances
01670044 StarterPac Installation with ACC Excellence XSR Precision Balances
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Numéro de catalogue. 01670024 Fournisseur METTLER TOLEDO™ Code fournisseur B99494130126
Il en reste null
Professional installation completed by a trained service technician will guarantee maximum performance and optimal use.
To achieve peak performance and compliance with quality systems and regulatory requirements, take the right steps at the beginning of equipment use. The Accuracy Calibration Certificate (ACC) is the new benchmark in the calibration of weighing equipment. Tests include repeatability, eccentricity and error of indication. A statement of measurement uncertainty is also included.
  • StarterPac includes: Professional installation performed by a trained and authorized service technician, installation confirmation/documentation, immediate deployment and use of your equipment, professionally trained operating personnel
  • IPac includes: Documented standard qualification and initial calibration, compliance with internal and external regulations, immediate deployment for quick, safe release to routine operation, and improved productivity with correct equipment set-up and initial user training.
  • EQPac includes: Regulatory conformance qualification documentation, professional qualification including test procedures, specification of weighing ranges and calibration, immediate deployment for quick, safe release to begin routine operation, improved productivity with correct equipment set-up and initial user training, cost savings through the use of a manufacturer developed qualification protocol.
Spécifications
For Use With (Equipment) Excellence XSR Precision Balances
Duration 1 Day
Product Type Balance Service
Service Offering Installation Qualification
Includes EQPac Regulated Balances with USP