missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Serum Vials, Clear
Made from premium Type I Borosilicate Glass, known for its durability and chemical resistance, this material is perfect for high-performance parenteral drug packaging.
$261.09 - $6,149.62
Specifications
| Color | Clear |
|---|---|
| Material | Borosilicate Glass (Type 1) |
| Pre-slit | No |
| Sterility | Non-sterile |
| Disposable | Single-use |
| Catalog Number | Mfr. No. | Volume (Metric) | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Volume (Metric) | Price | Quantity & Availability | |||||
|
02993451
|
Fisherbrand™
337377 |
50 mL |
Case of 130 Each for $261.09
|
|
|||||
|
02993453
|
Fisherbrand™
337379 |
100 mL |
Case of 88 Each for $267.50
|
|
|||||
|
02993454
|
Fisherbrand™
337396 |
2 mL |
Case of 4950 Each for $6,149.62
|
|
|||||
|
02993455
|
Fisherbrand™
337397 |
5 ml |
Case of 1200 Each for $1,318.79
|
|
|||||
|
02993458
|
Fisherbrand™
337401 |
10 mL |
Case of 800 Each for $1,042.12
|
|
|||||
|
02993459
|
Fisherbrand™
337404 |
20 mL |
Case of 576 Each for $833.68
|
|
|||||
Description
- ISO 9001:2015 Certified:
- Adhering to the highest international quality standards, ensuring consistent product excellence.
- Complete Batch Traceability:
- Full production traceability from raw materials to final product, ensuring accountability and quality control.
- Rigorous Quality Checks:
- Quality control at every stage of production, from raw material inspection to final packaging, ensuring product integrity.
Specifications
| Clear | |
| No | |
| Single-use | |
| Stoppers and Caps (Not Included) | |
| Autoclavable | |
| Serum Vial |
| Borosilicate Glass (Type 1) | |
| Non-sterile | |
| No | |
| No | |
| ISO 9001:2015 |