missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Serum Vials, Clear

Made from premium Type I Borosilicate Glass, known for its durability and chemical resistance, this material is perfect for high-performance parenteral drug packaging.

Supplier:  Fisherbrand™ 337396

 View more versions of this product

Catalog No. 02993454

Only null left

Description

Description

  • ISO 9001:2015 Certified:
  • Adhering to the highest international quality standards, ensuring consistent product excellence.
  • Complete Batch Traceability:
  • Full production traceability from raw materials to final product, ensuring accountability and quality control.
  • Rigorous Quality Checks:
  • Quality control at every stage of production, from raw material inspection to final packaging, ensuring product integrity.
Specifications

Chemical Identifiers

Clear
No
Single-use
Stoppers and Caps (Not Included)
Autoclavable
Serum Vial
Borosilicate Glass (Type 1)
Non-sterile
No
No
ISO 9001:2015

Specifications

Clear
Borosilicate Glass (Type 1)
Non-sterile
2 mL
4950
Tubular with blowback crimp neck finish
4950
No
ISO 9001:2015
Serum Vial
14.75 mm
No
Single-use
No
495
Stoppers and Caps (Not Included)
13 mm
Autoclavable
32 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions