missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Serum Vials, Clear

Catalog No. 02993455
Change view
Click to view available options
Volume (Metric):
2 mL
5 ml
10 mL
20 mL
50 mL
100 mL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
02993455 5 ml
02993451 50 mL
02993453 100 mL
02993454 2 mL
02993458 10 mL
02993459 20 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 02993455 Supplier Fisherbrand™ Supplier No. 337397
Only null left

Made from premium Type I Borosilicate Glass, known for its durability and chemical resistance, this material is perfect for high-performance parenteral drug packaging.

  • ISO 9001:2015 Certified:
  • Adhering to the highest international quality standards, ensuring consistent product excellence.
  • Complete Batch Traceability:
  • Full production traceability from raw materials to final product, ensuring accountability and quality control.
  • Rigorous Quality Checks:
  • Quality control at every stage of production, from raw material inspection to final packaging, ensuring product integrity.

Specifications

Color Clear
Diameter (Metric) Outer 21.75 mm
Material Borosilicate Glass (Type 1)
Pre-slit No
Sterility Non-sterile
Disposable Single-use
Volume (Metric) 5 ml
Graduated No
No. per Case 1200
No. per Pack 100
Style Tubular with blowback crimp neck finish
Closure Type Stoppers and Caps (Not Included)
Quantity 1200
Closure Size 20 mm
Marking Spot No
Autoclavable Autoclavable
Certifications/Compliance ISO 9001:2015
Height (Metric) 40 mm
Product Type Serum Vial
Show More Show Less