Learn More
Greiner Bio-One Serum No Additive Tubes
Numéro de catalogue. p-2770742
Change view
Click to view available options
Closure Color:
White with Black Ring
Tube Size:
13 x 75 mm
13 x 100 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Closure Color Tube Size
22040116 White with Black Ring 13 x 75 mm
22040163 White with Black Ring 13 x 100 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. 22040116 Fournisseur Greiner Bio-One Code fournisseur 454241
Il en reste null
With non-ridged pull caps
  • Use for discard tubes, immunohematology or viral marker testing
  • Clear as glass with equal or better performance
  • Molded to be virtually unbreakable, providing a better level of safety than glass
  • Tubes have an 18-month shelf life from the date of manufacture
  • Nonscrew Safety Pull Caps can be removed with a gentle pull
Spécifications
Material Plastic
Closure Type Non-ridged Pull Cap
Closure Color White with Black Ring
Tube Size 13 x 75 mm