missing translation for 'onlineSavingsMsg'
Learn More

Sertraline Hydrochloride Impurity Standard, British Pharmacopoeia (BP) Reference Standard, MilliporeSigma™ Supelco™

Catalog No. 111022961
Change view
Click to view available options
Quantity:
20 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111022961 20 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111022961 Supplier Merck Emd Millipore Supplier No. BP897

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets, has been developed and issued under the Authority of the issuing Pharmacopoeia.

Specifications

CAS 79559-97-0
Molecular Formula C17H17NCl2 · HCl
Linear Formula C17H17NCl2 · HCl
MDL Number MFCD00895772
Synonym Sertraline hydrochloride; (1 S,4 S)-4-(3,4-Dichlorophenyl)-1,2,3,4-tetrahydro-N-methyl-1-naphthalenamine hydrochloride
Quantity 20 mg
Formula Weight 342.69
Grade British Pharmacopoeia (BP) Reference Standard