missing translation for 'onlineSavingsMsg'
Learn More

Sertindole 95.0+%, TCI America™

Numéro de catalogue. S094210MG
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
S094210MG 10 mg
S094250MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. S094210MG Fournisseur TCI America Code fournisseur S094210MG
Il en reste null

Identifiants chimiques

CAS 106516-24-9
Molecular Formula C24H26ClFN4O
Molecular Weight (g/mol) 440.95
MDL Number MFCD00867749
InChI Key GZKLJWGUPQBVJQ-UHFFFAOYSA-N
Synonym 1-[2-[4-[5-Chloro-1-(4-fluorophenyl)-1H-indol-3-yl]-1-piperidinyl]ethyl]-2-imidazolidinone
PubChem CID 60149
ChEBI CHEBI:9122
IUPAC Name 1-(2-{4-[5-chloro-1-(4-fluorophenyl)-1H-indol-3-yl]piperidin-1-yl}ethyl)imidazolidin-2-one
SMILES FC1=CC=C(C=C1)N1C=C(C2CCN(CCN3CCNC3=O)CC2)C2=CC(Cl)=CC=C12

Spécifications

Melting Point 155°C
Color White
Quantity 10 mg
Formula Weight 440.95
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Sertindole
RTECSNumber NJ0621100
TSCA No
Recommended Storage Refrigerator