missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Sera-Mag™ Streptavidin Magnetic Bead, medium biotin binding capacity

Their cauliflower-like structure gives Sera-Mag beads high binding capacity. Sera-Mag Streptavidin beads contain covalently bound streptavidin and are available with low, medium, or high nominal biotin binding capacities. Choose the biotin-binding capacity needed to optimize your application.

$304.54 - $7,782.98

Spécifications

Concentration 1% solids (10mg/mL)
Content And Storage 2°C to 8°C
For Use With (Application) Immunoassays, cDNA Library Construction, Capture of Biotinylated Targets
Diameter (Metric) 1 μm
voir les spécifications

Produits 2
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantity & Availability  
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantity & Availability  
09981131
Afficher les documents Détails de la promotion
Cytiva
30152104010350
100 mL
chaque for $7,782.98
Il en reste null
 
09981129
Afficher les documents Détails de la promotion
Cytiva
30152104011150
1 mL
chaque for $304.54
Il en reste null
 
Description

Description

Biotin Binding Capacity of 3500 to 4500pmol/mg beads
Spécifications

Spécifications

1% solids (10mg/mL)
2°C to 8°C
Immunoassays, cDNA Library Construction, Capture of Biotinylated Targets
1 μm
Videos
FDS
Documents

Documents