missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Supelco™ Septa for Open Top Caps
For use with cut-out top caps and Wheaton vials.
$117.00 - $478.00
| Catalog Number | Mfr. No. | Material | Closure Size | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Material | Closure Size | Price | Quantity & Availability | ||||
|
111009517
|
MilliporeSigma™ Supelco™
Z127302100EA |
PTFE/Silicone | 20 mm |
Each for $478.00
|
|
||||
|
111009972
|
MilliporeSigma™ Supelco™
Z106518100EA |
PTFE/Silicone | 15 mm |
Each for $335.00
|
|
||||
|
111010856
|
MilliporeSigma™ Supelco™
Z106534100EA |
PTFE/Red Rubber | 15 mm |
Each for $166.00
|
|
||||
|
111011233
|
MilliporeSigma™ Supelco™
Z106526100EA |
PTFE/Styrene-butadiene Rubber | 13 mm |
Each for $117.00
|
|
||||