missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Septa Barrier

Catalog No. 111010919
Change view
Click to view available options
For Use With (Equipment):
20, 40 or 60 mL Vial
22 mL Vial
4 mL Vial
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
111010919 4 mL Vial
111010356 20, 40 or 60 mL Vial
111010833 22 mL Vial
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 111010919 Supplier MilliporeSigma™ Supelco™ Supplier No. 24883

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Septa barrier silicone bonded to aluminum

Specifications

Material Aluminum, Silicone
No. per Pack 100
Style Septum type Barrier
Thickness (English) 0.06 in.
Thickness (Metric) 11 mm
For Use With (Equipment) 4 mL Vial