missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Sephadex™ Ion Exchange Media

Separate molecules over broad molecular weight and pH ranges

Supplier:  Cytiva 17019001

 View more versions of this product

Catalog No. 45-000-038

Only null left
Explore available promotions

Description

Description

Range of weak and strong anionic (A) and cationic (C) exchangers. Fractionation range: A-25 and C-25, up to 30 000; A-50 and C-50: 30 000–100 000.

Very high capacities even at elevated ionic strengths
Very low non-specific adsorption
Wide range of applications at laboratory and process scales
Suitable for column or batch techniques
Simple and economical to use

DEAE Sephadex

  • Weak and strong anion exchanger
  • pH range 2-9

QAE Sephadex
  • Strong anion exchanger
  • pH range: 2-12
CM Sephadex
  • Weak cation exchanger
  • pH range: 6-10
SP Sephadex
  • Strong cation exchanger
  • pH range: 4-13

Specifications

Specifications

100 g
Column or Batch Techniques
Solid Spherical Bead
2 to 5.9 in.
Diethyl- (2-hydroxy-propyl) Aminoethyl
2 to 12
Up to 30,000
40 to 120 μm in dry form
Sephadex Ion Exchange Media
Separating and purifying Proteins, Polypeptides, Nucleic Acids and other charged biomolecules by using column or batch techniques
5 to 15cm
QAE-Sephadex A-25
Cross-linked Dextran
4° to 30°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions