missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Sephadex™ Ion Exchange Media

Catalog No. 45000035
Change view
Click to view available options
Quantity:
100 g
500 g
Material:
CM-Sephadex C-25
CM-Sephadex C-50
DEAE-Sephadex A-25
DEAE-Sephadex A-50
QAE-Sephadex A-25
QAE-Sephadex A-50
SP-Sephadex C-25
MWCO:
30,000 to 100,000
Up to 30,000
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Material MWCO
45000035 500 g DEAE-Sephadex A-25 Up to 30,000
45000040 100 g QAE-Sephadex A-50 30,000 to 100,000
45000037 500 g DEAE-Sephadex A-50 30,000 to 100,000
45000034 100 g DEAE-Sephadex A-25 Up to 30,000
45000036 100 g DEAE-Sephadex A-50 30,000 to 100,000
45000038 100 g QAE-Sephadex A-25 Up to 30,000
45000041 100 g CM-Sephadex C-25 Up to 30,000
45000042 500 g CM-Sephadex C-25 Up to 30,000
45000044 500 g CM-Sephadex C-50 30,000 to 100,000
45000043 100 g CM-Sephadex C-50 30,000 to 100,000
45000045 100 g SP-Sephadex C-25 Up to 30,000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. 45000035 Supplier Cytiva Supplier No. 17017002
Only null left

Separate molecules over broad molecular weight and pH ranges

Range of weak and strong anionic (A) and cationic (C) exchangers. Fractionation range: A-25 and C-25, up to 30 000; A-50 and C-50: 30 000–100 000.

Very high capacities even at elevated ionic strengths
Very low non-specific adsorption
Wide range of applications at laboratory and process scales
Suitable for column or batch techniques
Simple and economical to use

DEAE Sephadex

  • Weak and strong anion exchanger
  • pH range 2-9

QAE Sephadex
  • Strong anion exchanger
  • pH range: 2-12
CM Sephadex
  • Weak cation exchanger
  • pH range: 6-10
SP Sephadex
  • Strong cation exchanger
  • pH range: 4-13

Specifications

Quantity 500 g
Particle Size 40 to 120 μm in dry form
For Use With (Equipment) Column or Batch Techniques
Product Type Sephadex Ion Exchange Media
Format Solid Spherical Bead
Diameter (Metric) Inner 5cm
For Use With (Application) Separating and purifying Proteins, Polypeptides, Nucleic Acids and other charged biomolecules by using column or batch techniques
Height (English) 2 to 5.9 in.
Height (Metric) 5 to 15cm
Ligand Type Diethylaminoethyl
Material DEAE-Sephadex A-25
pH Range 2 to 9
Support Type Cross-linked Dextran
MWCO Up to 30,000
Content And Storage 4° to 30°C
Show More Show Less