missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Sephadex™ G50 Medium Media

Catalog No. 45000018
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45000018 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45000018 Supplier Cytiva Supplier No. 17004301

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Suitable for gel filtration

  • Particle size range of wet beads: 100 to 300μm
  • Fractionation range (Mr), globular proteins: 1500 to 30,000

Specifications

Quantity 100 g
Flow Rate < Darcy´s Law
Particle Size 100 to 300 μm in wet form, 50 to 150 μm in dry form
Bed Volume 9 to 11mg/g
Product Type Sephadex G50 Media
Format Solid Bead
Diameter (Metric) Inner 5cm
For Use With (Application) For the separation of molecules Mr >30 000 from molecules Mr <1500 such as labeled protein or DNA from unconjugated dyesFor removeing small nucleotides from longer chain nucleic acids
Height (English) 3.93 in.
Height (Metric) 10cm
Max. Pressure min 150 cm/h, bed height 10 cm, column 5 cm i.d.
pH Range 2 to 10
Support Type Cross-linked Dextran
Sample Type DNA
Content And Storage 4 to 30°C
Show More Show Less