missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Sephadex™ G50 Fine Media
Description
- Particle size range 40 to 160μm
Specifications
Specifications
| Quantity | 100 g |
| Flow Rate | < Darcy´s Law |
| Particle Size | 34 to 208 μm in wet form, 20 to 80 μm in dry form |
| Bed Volume | 9 to 11mg/g |
| Product Type | Sephadex G50 Media |
| Format | Solid Bead |
| Diameter (Metric) Inner | 5cm |
| For Use With (Application) | For the separation of molecules Mr >30 000 from molecules Mr <1500 such as labeled protein or DNA from unconjugated dyesFor removeing small nucleotides from longer chain nucleic acids |
| Height (English) | 3.93 in. |
| Height (Metric) | 10cm |
| Show More |