missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Sephadex™ G50 Fine Media

Catalog No. 45000017
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45000017 500 g
45000016 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45000017 Supplier Cytiva Supplier No. 17004202

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Suitable for gel filtration

  • Particle size range 40 to 160μm

Specifications

Quantity 500 g
Flow Rate < Darcy´s Law
Particle Size 34 to 208 μm in wet form, 20 to 80 μm in dry form
Bed Volume 9 to 11mg/g
Product Type Sephadex G50 Media
Format Solid Bead
Diameter (Metric) Inner 5cm
For Use With (Application) For the separation of molecules Mr >30 000 from molecules Mr <1500 such as labeled protein or DNA from unconjugated dyesFor removeing small nucleotides from longer chain nucleic acids
Height (English) 3.93 in.
Height (Metric) 10cm
Max. Pressure min 150 cm/h, bed height 10 cm, column 5 cm i.d.
pH Range 2 to 10
Support Type Cross-linked Dextran
Sample Type DNA
Content And Storage 4 to 30°C
Show More Show Less