missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Sephadex™ G10 Media

Catalog No. 45000012
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45000012 500 g
45000011 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45000012 Supplier Cytiva Supplier No. 17001002

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Gel filtration medium for desalting and buffer exchange of peptides and small biomolecules

  • Beads behave as rigid spheres obeying Darcy's Law for purposes of determining maximum flowrate and maximum operating pressure
  • Working pH range 2 to 13
  • Particle size range 55 to 165μm
  • Bed volume in distilled water 2-3mL/g
  • Exclusionation limit: 7 x 102 dextrans, 7 x 102 globular proteins

Specifications

Quantity 500 g
Flow Rate < Darcy´s Law
Particle Size 55 to 165 μm in wet form, 40 to 120 μm in dry form
Bed Volume 2 to 3mL/g in Distilled Water
Product Type Sephadex G10 Media
Format Solid Bead
Diameter (Metric) Inner 2.6cm
For Use With (Application) Desalting and Buffer exchange of Peptides and small biomolecules >700 M.W.
Height (English) 11.81 in.
Height (Metric) 30cm
Max. Pressure 24-49 cm/h, pressure drop cm H2O/bed height = 2, bed height 30 cm, 2.6 cm i.d
pH Range 2 to 13
Support Type Cross-linked Dextran
Sample Type Peptides, Small Biomolecules
Content And Storage 4 to 30°C
Show More Show Less