missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Sephadex™ G-25 Media
Description
Established medium for desalting and buffer exchange
- Gel filtration medium
- Available in coarse, medium, fine and superfine grades
- All grades satisfy process-scale chromatography requirements
- Available in lab packs or prepacked columns
Specifications:
- Working pH range: 2 to 13
- Bed volume (in distilled water): 4 to 6mL/g
- Fractionation range dextrans/globular proteins: 1× 102 to 5 × 103 / 1 × 103 to 5 × 103 Mr
- Max. flowrate/operating pressure: the beads behave as rigid spheres obeying Darcy's Law
Specifications
Specifications
| Quantity | 500 g |
| Flow Rate | >47 cm/hr. |
| Particle Size | 17 to 132 μm (Wet Form), >20 μm (Dry Form) |
| Bed Volume | 4 to 6mL/g in Distilled Water |
| Product Type | Sephadex G-25 Media |
| Format | Solid Spherical Bead |
| Diameter (Metric) Inner | 2.6cm |
| For Use With (Application) | Desalting and Buffer exchange and removal free label in industrial applications |
| Height (English) | 11.81 in. |
| Height (Metric) | 30cm |
| Show More |