missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Sephacryl™ S-500 High Resolution Media

Catalog No. p-4895772
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
150 mL
750 mL
Catalog No. Quantity
45002053 150 mL
45002054 750 mL
2 options

Catalog No. 45002053

Supplier: Cytiva 17061310

Only null left

Hydrophilic, rigid allyl dextran/bisacrylamide matrix for fast and reproducible purification of proteins, polysaccharides and other macromolecules

Chromatography media offer good recovery and low nonspecific adsorption. Enable gel filtration at laboratory and industrial scale, and may be purchased in prepacked columns.

  • For separating polysaccharides, macromolecules with extended structures, and small plasmids
  • Excellent purification over a wide molecular weight range
  • High reproducibility due to long-term chemical and physical stability
  • High flow rates and recoveries
  • Well-suited to industrial-scale use
  • Storage conditions: 20% ethanol; 4° to 30°C
  • Available in a variety of selectivities and volumes

Chemical Stability (stable in all common buffers):

  • 1M acetic acid, 8M urea
  • 6M guanidine-HCl, 1% SDS
  • 2M NaCl, 24% ethanol
  • 30% propanol, 30% acetonitrile
  • 0.5M NaOH (for cleaning)
pH Stability
  • 3 to 11 (long-term and working)
  • 2 to 13 (short-term)
Physical Stability:
  • Negligible volume variation due to changes in pH or ionic strength
  • Autoclavability: At 121°C, pH 7 for 30 minutes (gel only)
  • Antimicrobial agent: 20% ethanol

Specifications

Quantity 150 mL
Flow Rate >125 cm/hr.
Particle Size 25 to 75 μm
Bed Volume 19 to 230L
Product Type Sephacryl HR Media
Format Solid Spherical Bead
Diameter (Metric) Inner 40 to 140cm
For Use With (Application) Allow fast and reproducible purification of proteins, polysaccharides and other macromolecules by gel filtration at laborato ry and industrial scale
Height (English) 5.9 in.
Height (Metric) 15cm
Max. Pressure 100kPa
pH Range 3 to 11
Support Type Allyl Dextran and N, N'-methylenebisacrylamide
Sample Type Polysaccharides, macromolecules and small plasmids
Content And Storage 4° to 30°C
Show More Show Less