missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EKF HemataCHEK™ Hematocrit Control
Ideal for use with HemataSTAT™ and other microhematocrit centrifuges
$648.14 - $729.18
Specifications
| Content And Storage | 40°C to 80°C |
|---|---|
| Format | Vial |
| For Use With (Equipment) | Microhematocrit centrifuge |
| Type | HemataCHEK™ Hematology Reference Control |
| Intended Use | For monitoring the Hematocrit determination |
| Catalog Number | Mfr. No. | Level | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Level | Price | Quantity & Availability | |||||
|
23550105
|
EKF
300101 |
Normal |
Pack of 6 Each for $727.89
|
|
|||||
|
23550107
|
EKF
300103 |
Low and Normal |
Pack of 6 Each for $648.14
|
|
|||||
|
23550106
|
EKF
300102 |
Low, Normal and High |
Pack of 6 Each for $729.18
|
|
|||||
|
23550109
|
EKF
SEP300105 |
Low |
Pack of 6 Each for $727.89
|
|
|||||
|
23550110
|
EKF
300107 |
High |
Pack of 6 Each for $648.14
|
|
|||||
|
23550108
|
EKF
300104 |
Normal, High |
Pack of 6 Each for $648.14
|
|
|||||
Description
- Assayed for HemataSTAT II and other microhematocrit centrifuges
- Comes in Low, Normal and High levels
Order Info
Box of 6 vials; 2.5mL per vial
Specifications
| 40°C to 80°C | |
| Microhematocrit centrifuge | |
| For monitoring the Hematocrit determination | |
| Human Blood |
| Vial | |
| HemataCHEK™ Hematology Reference Control | |
| 6 Pack | |
| 31 Days Open Vial |
Safety and Handling
ShelfLife : 2 Years