missing translation for 'onlineSavingsMsg'
Learn More

EKF HemataCHEK™ Hematocrit Control

Catalog No. 23550106
Change view
Click to view available options
Level:
High
Low
Low, Normal
Low, Normal, High
Normal
Normal, High
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Level
23550106 Low, Normal, High
23550105 Normal
23550107 Low, Normal
23550109 Low
23550110 High
23550108 Normal, High
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 23550106 Supplier EKF Supplier No. 300102

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ideal for use with HemataSTAT™ and other microhematocrit centrifuges

  • Assayed for HemataSTAT II and other microhematocrit centrifuges
  • Comes in Low, Normal and High levels

Order Info

Box of 6 vials; 2.5mL per vial

Specifications

Content And Storage 40°C to 80°C
Format Vial
For Use With (Equipment) Microhematocrit centrifuge
Type HemataCHEK™ Hematology Reference Control
Intended Use For monitoring the Hematocrit determination
Level Low, Normal, High
Quantity 6 Pack
Source Human Blood
Stability 31 Days Open Vial