missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EKF HemataCHEK™ Hematocrit Control
Ideal for use with HemataSTAT™ and other microhematocrit centrifuges
Supplier: EKF 300103
Description
- Assayed for HemataSTAT II and other microhematocrit centrifuges
- Comes in Low, Normal and High levels
Order Info
Box of 6 vials; 2.5mL per vial
Chemical Identifiers
| 40°C to 80°C | |
| Microhematocrit centrifuge | |
| For monitoring the Hematocrit determination | |
| Human Blood |
| Vial | |
| HemataCHEK™ Hematology Reference Control | |
| 6 Pack | |
| 31 Days Open Vial |
Specifications
| 40°C to 80°C | |
| Microhematocrit centrifuge | |
| For monitoring the Hematocrit determination | |
| 6 Pack | |
| 31 Days Open Vial |
| Vial | |
| HemataCHEK™ Hematology Reference Control | |
| Low and Normal | |
| Human Blood |
Safety and Handling
ShelfLife : 2 Years