missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EKF HemataCHEK™ Hematocrit Control
Description
- Assayed for HemataSTAT II and other microhematocrit centrifuges
- Comes in Low, Normal and High levels
Order Info
Box of 6 vials; 2.5mL per vial
Specifications
Specifications
| Content And Storage | 40°C to 80°C |
| Format | Vial |
| For Use With (Equipment) | Microhematocrit centrifuge |
| Type | HemataCHEK™ Hematology Reference Control |
| Intended Use | For monitoring the Hematocrit determination |
| Level | Low |
| Quantity | 6 Pack |
| Source | Human Blood |
| Stability | 31 Days Open Vial |