missing translation for 'onlineSavingsMsg'
Learn More

EKF Disposable Tube Holders

Catalog No. 23550111
Change view
Click to view available options
Color:
Clear
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
23550111 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23550111 Supplier EKF Supplier No. SEP230100
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For use with HemataSTAT™ or Creamatocrit Plus™ centrifuges

  • Clear, disposable tube holders

Specifications

Disposable Single-use
Color Clear
For Use With (Equipment) HemataSTAT™ or Creamatocrit Plus™ centrifuges
Product Type Tube Holder