Learn More
Thermo Scientific™ Sensors and Replacement Batteries for Thermo Scientific™ Ultra-Low Temperature Freezers
Catalog No. 400159
Change view
Click to view available options
Type:
Replacement Back-Up Battery
Catalog No. Type
400159 Replacement Back-Up Battery
1 options
Catalog No. 400159 Supplier Thermo Scientific™ Supplier No. 400159
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Use these Thermo Scientific™ Sensors and Replacement Batteries with our ultra-low temperature freezers.
Specifications
Type Replacement Back-Up Battery
For Use With (Equipment) Forma™900, Forma™ 8600 Series, Forma™ 88000, TSD Series, TSUSeries
The low battery alarm is displaying on my ULT (Ultra-Low Temperature) freezer. Can I buy a new battery?
You can order a new battery (Cat. No. 400159) online.