missing translation for 'onlineSavingsMsg'
Learn More

Sensidyne™ Gilian™ AirCon-2™ Constant-Flow Samplers

Catalog No. 19087543
Encompass
Change view
Click to view available options
Flow Rate:
2 to 3 L/min.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Flow Rate
19087543 2 to 3 L/min.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19087543 Supplier Sensidyne™ Supplier No. 801012100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

AC models come with removable AC line cord, independent mast stand, tubing, and instruction manual. DC model comes with mast stand, tubing, and instruction manual.

Feature 2 to 30L/min. constant flow range

Portable high-volume samplers use filter cassette (sold separately) to sample for particulates, dust, and chemical hazards. Constant-flow capabilities from 2 to 30L/min. at pressures to 7psi. Choice of basic nontimer model with AC power configuration or programmable models with AC or DC power configurations. Programmable timing functions include run, delay, start/stop, intermittent run, and hold times. Adjustable mast stand extends filter cassette into breathing zone and keeps it independent of sampler, allowing vibration-free collection of particulates.

Order Info

Requires battery pack for operation (order separately, below). Battery packs are rated at four hours and are stackable for up to eight hours of sampling. Measure 5 1/4L x 7 1/2W x 10 1/4 in.H (13.3 x 19.1 x 26cm). Net weight: 12 lb. (5.4kg).

Specifications

Includes Removable AC Line Cord (AC Models), Independent Mast Stand (AC and DC Models), Tubing (AC and DC Models), Instruction Manual (AC and DC Models)
Certifications/Compliance NIOSH, OSHA
For Use With (Application) Use filter cassette to sample for particulates, dust, and chemical hazards
Dimensions (L x W x H) 5.25 in. x 7.25 in. x 10.25 in. (13.3 cm x 13.3 cm x 26 cm)
Overall Length (English) 5.25 in.
Overall Length (Metric) 13.3 cm
Weight (English) 12 lb.
Product Type Programmable DC
Flow Rate 2 to 3 L/min.
Weight (Metric) 5.4 kg
Overall Height (English) 10.25 in.
Overall Height (Metric) 26 cm
Overall Width (English) 7.25 in.
Overall Width (Metric) 13.3 cm
Operating Temperature Range (English) -4° to 113°F
Operating Temperature Range (Metric) -4° to 45°C
Show More Show Less