missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sennoside A, ≥96% (HPLC), MilliporeSigma™ Supelco™
Description
Featured Industries: Food and Beverages
Specifications
Specifications
| CAS | 81-27-6 |
| Physical Form | Neat |
| Linear Formula | C42H38O20 |
| MDL Number | MFCD00151527 |
| Type | Analytical Standard |
| Quantity | 25 mg |
| Formula Weight | 862.74 |
| Percent Purity | ≥96% (HPLC) |
| Grade | Analytical Standard |