missing translation for 'onlineSavingsMsg'
Learn More

Semi-Volatile Acid Surrogate, SPEX CertiPrep™

Catalog No. CLP9075SA5
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CLP9075SA5 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CLP9075SA5 Supplier SPEX CertiPrep Group LLC Supplier No. CLP9075SA5

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
2,4,6-Tribromophenol118-79-67500μg/mL
2-Chlorophenol-3,4,5,6-d493951-73-67500μg/mL
2-Fluorophenol367-12-47500μg/mL
Phenol-d613127-88-37500μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Chemical Name or Material Semi-Volatile Acid Surrogate
Packaging 5mL amber prescored glass ampule
CAS 118-79-6, 93951-73-6, 367-12-4, 13127-88-3
Concentration or Composition Notes Certificate of Analysis reports target analytes to a ±2% accuracy and correct for raw material purity.
Analyte or Component Names 2,4,6-Tribromophenol
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
Type Certified Reference Material
Element 2,4,6-Tribromophenol
Solvent or Matrix Methanol
Quantity 5 mL
Concentration or Composition (by Analyte or Components) 7500μg/mL
Show More Show Less

Product intended for laboratory use only