missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Selleger's Stain, Reagents
Spécifications
Spécifications
| Color | Brown |
| Form | Liquid |
| Molecular Formula | CaH8N2O10 |
| Packaging | Glass Bottle |
| Packaging Type | Glass Bottle |
| InChI Key | MWGCGFYACDTFSB-UHFFFAOYSA-N |
| SMILES | O.[Ca++].[O-][N+]([O-])=O |
| IUPAC Name | calcium bis(nitric acid) tetrahydrate |
| Molecular Weight (g/mol) | 236.15 |
| Quantity | 30 mL |
| Afficher plus de résultats |