Learn More
GA International Self-Laminating Calibration Labels - ""Inspected"" 1"" x 2.125"" / 25.4mm x 54mm Rectangle Black text on Blue From -80C to +93C (-112F to +199.4F)
Numéro de catalogue. 5000571822
Encompass
Self-laminating, writable, calibration labels designed for equipment that is calibrated or inspected regularly. The labels can be inscribed using permanent ink markers. The clear laminate is applied over the label protecting the written information. CalTAG™ labels can withstand a wide range of temperatures (-80C to +72C / -112F to +161F), abrasion and exposure to harsh chemicals, such as ethanol, acetone, isopropanol, thinners, detergents and surface sanitizing wipes.The self-laminating calibration labels are ideal for labeling regularly inspected medical equipment, equipment that has undergone preventative maintenance, that require calibration prior to use, or that are out of order and must not be used. Reagent calibration labels are also available for time-sensitive solutions prepared in the lab. Calibration labels customization is also available to include the companys logos and information.
Numéro de catalogue. 5000571822 Fournisseur GA International Code fournisseur CALA0151R
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Self-laminating, writable, calibration labels designed for equipment that is calibrated or inspected regularly. The labels can be inscribed using permanent ink markers. The clear laminate is applied over the label protecting the written information. CalTAG™ labels can withstand a wide range of temperatures (-80C to +72C / -112F to +161F), abrasion and exposure to harsh chemicals, such as ethanol, acetone, isopropanol, thinners, detergents and surface sanitizing wipes.The self-laminating calibration labels are ideal for labeling regularly inspected medical equipment, equipment that has undergone preventative maintenance, that require calibration prior to use, or that are out of order and must not be used. Reagent calibration labels are also available for time-sensitive solutions prepared in the lab. Calibration labels customization is also available to include the companys logos and information.