missing translation for 'onlineSavingsMsg'
Learn More

Selenium AA Standard, Certified, 1000ppm ±10ppm (1mL = 1mg Se), LabChem™

Supplier:  LabChem LC223701

 View more versions of this product

Catalog No. LC223701


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Selenium AA Standard
Poly Bottle
95.9,4,0.1
Liquid
1000 ppm ±10 ppm (1mL = 1mg Se)
MFCD00134090 MFCD00011224
Colorless
Soluble in water
[Se]
78.97
6326970
78.96
1000ppm (1mL = 1mg Se)
7782-49-2 , 7782-49-2 , 7732-18-5
Passes Test
Se
Se
UN3264
105°C
BUGBHKTXTAQXES-UHFFFAOYSA-N
selenium
500 mL
CHEBI:27568
Certified

Chemical Identifiers

7782-49-2
78.97
BUGBHKTXTAQXES-UHFFFAOYSA-N
CHEBI:27568
[Se]
Se
MFCD00134090 MFCD00011224
6326970
selenium
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
May be corrosive to metals.
Causes severe skin burns and eye damage.

GHS P Statement
Keep only in original container.
Do not breathe mist, vapors, spray.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Absorb spillage to prevent material damage.
Store locked up.
Store in corrosive resistant container with a resistant inner liner.
Dispose of contents/container to comply with local, state and federal regulations.

Danger

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions