missing translation for 'onlineSavingsMsg'
Learn More

Selectophore™ Tetrahydrofuran, For ion-selective electrodes, ≥99.5%, MilliporeSigma™ Supelco™

Catalog No. 111018529
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018529 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018529 Supplier Merck Emd Millipore Supplier No. 87369500ML
Only null left

Tetrahydrofuran (THF), an organic compound which belongs to saturated cyclic ether group. It is mainly used as solvent for PVC and organoalkali compounds. It is also used in lithium batteries.

Chemical Identifiers

MDL Number MFCD00005356
Synonym THF; Butylene oxide; Oxolane; Tetramethylene oxide

Specifications

Melting Point -108°C
CAS 109-99-9
Density 0.889 g/mL (at 25°C)
Boiling Point 65°C to 67°C
Flash Point -17°C
Ignition Point 610°F
Linear Formula C4H8 O
Quantity 500 mL
Solubility Information Water: soluble
Vapor Density 2.5 (vs. air)
Vapor Pressure 114 mmHg (15°C)
Formula Weight 72.11
Percent Purity ≥99.5% (GC); ≥99.5%
Grade For ion-selective electrodes
Product Line Selectophore
Show More Show Less
RTECSNumber LU5950000

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH