missing translation for 'onlineSavingsMsg'
Learn More

Selectophore™ Oleic acid, ≥99.0%, MilliporeSigma™ Supelco™

Catalog No. 111012509
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012509 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012509 Supplier Merck Emd Millipore Supplier No. 055085MLF

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Visit our Sensor Applications portal to learn more.

Chemical Identifiers

MDL Number MFCD00064242
Synonym cis-9-Octadecenoic acid; Elainic acid

Specifications

Melting Point 13°C to 14°C
Density 0.89 g/mL (at 25°C)
CAS 112-80-1
Boiling Point 194°C to 195°C/1.2 mmHg
Flash Point >113°C
Linear Formula CH3(CH2)7 CH=CH(CH2)7 COOH
Quantity 5 mL
Formula Weight 282.46
Vapor Pressure 1 mmHg (176°C)
Percent Purity ≥99.0% (GC); ≥99.0%
Grade Selectophore
Physical Form Liquid
Product Line Selectophore
Show More Show Less
ShelfLife Limited shelf life, expiry date on the label
RTECSNumber RG2275000

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH