missing translation for 'onlineSavingsMsg'
Learn More

Selectophore™ Lithium Ionophore VIII, Function Tested, MilliporeSigma™ Supelco™

Catalog No. 111012954
Change view
Click to view available options
Quantity:
50 mg
Packaging:
Bottomless Glass Bottle with Fused Cone
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
111012954 50 mg Bottomless Glass Bottle with Fused Cone
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012954 Supplier Merck Emd Millipore Supplier No. 6257150MGF
Only null left

Lipophilic, neutral ionophore for lithium ion-selective solvent polymeric membrane electrodes

Chemical Identifiers

CAS 133338-85-9
Molecular Formula C48H83N3O6
Molecular Weight (g/mol) 798.21
MDL Number MFCD00467019
InChI Key QSTVDAZAPLNMPD-UHFFFAOYSA-N
Synonym N,N,N′,N′,N′′,N′′-Hexacyclohexyl-4,4 ′,4 ′′-propylidynetris(3-oxabutyramide)
IUPAC Name N,N-dicyclohexyl-2-(2,2-di{[(dicyclohexylcarbamoyl)methoxy]methyl}butoxy)acetamide
SMILES CCC(COCC(=O)N(C1CCCCC1)C1CCCCC1)(COCC(=O)N(C1CCCCC1)C1CCCCC1)COCC(=O)N(C1CCCCC1)C1CCCCC1

Specifications

Packaging Bottomless Glass Bottle with Fused Cone
Linear Formula C48H83N3O6
Quantity 50 mg
Formula Weight 798.19
Grade Selectophore
Product Line Selectophore

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH