missing translation for 'onlineSavingsMsg'
Learn More

Selectophore™ Dipentyl phthalate, 99.0%, MilliporeSigma™ Supelco™

Catalog No. 111019504
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019504 1 mL
111018346 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111019504 Supplier Merck Emd Millipore Supplier No. 801541ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Plasticizer for PVC membrane electrodes

Chemical Identifiers

CAS 131-18-0
Molecular Formula C18H26O4
Molecular Weight (g/mol) 306.40
MDL Number MFCD00041934
InChI Key IPKKHRVROFYTEK-UHFFFAOYSA-N
IUPAC Name 1,2-dipentyl benzene-1,2-dicarboxylate
SMILES CCCCCOC(=O)C1=CC=CC=C1C(=O)OCCCCC

Specifications

Density 1.025 g/mL (at 20°C)
Linear Formula C18H26O4
Refractive Index n20/D 1.490
Quantity 1 mL
Formula Weight 306.40
Percent Purity ≥99.0% (GC)
Grade Selectophore
Product Line Selectophore
RTECSNumber TI1930000

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH