missing translation for 'onlineSavingsMsg'
Learn More

Selectophore™ Dichloromethane, 99.5%, MilliporeSigma™ Supelco™

$132.52 - $286.04

Specifications

Description Solvent for preparation of membranes
CAS 75-09-2
Product Line Selectophore
Ignition Point 1223°F
Solution Contains ˜0.005% amylene as stabilizer
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
111019812
View Documents
Merck Emd Millipore
66742100ML
100 mL
Each for $132.52
Only null left
 
111018930
View Documents
Merck Emd Millipore
66742500ML
500 mL
Each for $286.04
Only null left
 
Description

Description

Specifications

Specifications

Solvent for preparation of membranes
Selectophore
˜0.005% amylene as stabilizer
2.9 (vs. air)
CH2 Cl2
-97°C
Ion Selective Electrode
Organic Standard
≥99.5% (GC)
75-09-2
1223°F
24.45 psi (55°C)
1.324-1.326 g/mL (at 20°C)
MFCD00000881
39.8°C to 40°C
Methylene chloride
84.93
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH