missing translation for 'onlineSavingsMsg'
Learn More

Selectophore™ Dichloromethane, 99.5%, MilliporeSigma™ Supelco™

Catalog No. 111019812
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019812 100 mL
111018930 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111019812 Supplier Merck Emd Millipore Supplier No. 66742100ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Description Solvent for preparation of membranes
CAS 75-09-2
Product Line Selectophore
Ignition Point 1223°F
Solution Contains ˜0.005% amylene as stabilizer
Vapor Pressure 24.45 psi (55°C)
Vapor Density 2.9 (vs. air)
Density 1.324-1.326 g/mL (at 20°C)
Linear Formula CH2 Cl2
MDL Number MFCD00000881
Melting Point -97°C
Boiling Point 39.8°C to 40°C
For Use With (Equipment) Ion Selective Electrode
Synonym Methylene chloride
Type Organic Standard
Quantity 100 mL
Formula Weight 84.93
Percent Purity ≥99.5% (GC)
Show More Show Less

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH