missing translation for 'onlineSavingsMsg'
Learn More

Selectophore™ Chromoionophore III, MilliporeSigma™ Supelco™

Numéro de catalogue. 111013189
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
111013189 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 111013189 Fournisseur Merck Emd Millipore Code fournisseur 2708810MGF
Il en reste null

Chromoionophore III (ETH 5350) is a pH sensitive dye, and is ratiometric that is the fluorescence spectrum has two peaks, and the ratio between these peaks changes according to the changing pH levels.

Identifiants chimiques

MDL Number MFCD00213937
Synonym 9-(Diethylamino)-5-[(2-octyldecyl)imino]benzo[a]phenoxazine; ETH 5350

Spécifications

CAS 149683-18-1
Packaging Bottomless Glass Bottle with Inserted Fused Cone
Linear Formula C38H55N3 O
Quantity 10 mg
Formula Weight 569.86
Grade Selectophore
Product Line Selectophore

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH