missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Selecto™ Preparative and Classic Column Chromatography Silica
Description
- pH of 7
- Surface area of 500 to 600m2/g
Spécifications
Spécifications
| Particle Size | 63 to 200 μm |
| pH | 7 |
| Quantity | 25 kg |
| Material | Silica Gel |