missing translation for 'onlineSavingsMsg'
Learn More

SEKISUI Diagnostics™ OSOM™ Strep A Test Kit

Catalog No. 22071059
Change view
Click to view available options
Detectable Analytes:
Streptococci (Group A)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Detectable Analytes
22071059 Streptococci (Group A)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22071059 Supplier SEKISUI Diagnostics™ Supplier No. SEK 141

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

OSOM™ Strep A Test Kit provides fast results for better treatment—aids in diagnosis of Strep A, preventing the unnecessary use of antibiotics.

  • Simple-to-use - get easy to read results in four easy steps, with no need for additional equipment
  • Trust your results - high sensitivity and specificity with results in just 5 minutes
  • Easy-to-read two-color results - blue test line and red control line
  • Collect and detect - qualitative detection of Group A Streptococci antigen from throat swabs, can also confirm presumptive Group A Streptococcal colonies recovered from cultures less than 72 hours old
  • QC Inside™ - two extra tests included no additional charge to run external quality control testing for more confidence in your results
  • Made in the USA

Specifications

Certifications/Compliance CE Marked
Content And Storage Room Temperature (15°C to 30°C, 59°F to 86°F)
Control Sets Included
CPT Code 87880QW
Clia Complexity Waived, Moderate
Detectable Analytes Streptococci (Group A)
Format Manual Dipstick
Sample Type Throat Swab
Sensitivity 96%
Type Rapid Test
Includes 50 test sticks, 50 test tubes, 50 sterile swabs, 1 vial of Reagent 1, 1 vial of Reagent 2, 1 positive control, 1 negative control, 1 instructions for use (IFU), product insert. NOTE: Two extra test sticks are included in the kit for external QC testing.
Packaging Type Test Kit
Specificity 97.8%
Test Time 5 min.
Analysis Time 5 min.
Show More Show Less