missing translation for 'onlineSavingsMsg'
Learn More

Secologanin, ≥88% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111013424
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013424 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013424 Supplier Merck Emd Millipore Supplier No. 507415MGF
Only null left

Specifications

CAS 19351-63-4
Physical Form Neat
Linear Formula C17H24O10
MDL Number MFCD16661163
Type Analytical Standard
Quantity 5 mg
Formula Weight 388.37
Percent Purity ≥88% (HPLC)
Grade Analytical Standard