missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SEC23IP Polyclonal Antibody
GREENER_CHOICE

Numéro de catalogue. PIPA558378
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
PIPA558378 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. PIPA558378 Fournisseur Invitrogen™ Code fournisseur PA558378
Il en reste null

Rabbit Polyclonal Antibody

Immunogen sequence: HLELKESLSR MGSDLKQGFI SSLKSAWQTL NEFARAHTSS TQLQEELEKV ANQIKEEEEK QVVEAEKVVE SPDFSKDEDY LGKVGML Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 98%.

SEC23IP is involved in protein transport from the endoplasmic reticulum to the Golgi apparatus. The protein encoded by this gene was identified by its interaction with a mouse protein similar to yeast Sec23p, an essential component of the COPII. This protein shares significant similarity with phospholipid-modifying proteins, especially phosphatidic acid preferring-phospholipase A1. Overexpression of this protein has been shown to cause disorganization of the endoplasmic reticulum-Golgi intermediate compartment and Golgi apparatus, which suggests its role in the early secretory pathway.

Spécifications

Antigen SEC23IP
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SEC23IP
Gene Accession No. Q6NZC7, Q9Y6Y8
Gene Alias D7Ertd373e; MSTP053; P125; P125A; SEC23 interacting protein; SEC23-interacting protein; Sec23-interacting protein p125; SEC23-interacting protein-like protein; SEC23IP
Gene Symbols SEC23IP
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SEC23IP. Recombinant protein control fragment (Product #RP-97033).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11196, 207352, 309010
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Afficher plus de résultats Afficher moins de résultats