missing translation for 'onlineSavingsMsg'
Learn More

sec-Butanol, 99%, Extra Pure

Catalog No. AC107700100
Change view
Click to view available options
Quantity:
1 L
2.5 L
10 L
Packaging:
Glass Bottle
Plastic Drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC107700100 10 L Plastic Drum
AC107700010 1 L Glass Bottle
AC107700025 2.5 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC107700100 Supplier Thermo Scientific Chemicals Supplier No. 107700100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 78-92-2 | C4H10O | 74.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 78-92-2
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.12
MDL Number MFCD00004569
InChI Key BTANRVKWQNVYAZ-UHFFFAOYNA-N
Synonym 2-butanol, sec-butanol, sec-butyl alcohol, 2-hydroxybutane, 2-butyl alcohol, s-butyl alcohol, butylene hydrate, s-butanol, 1-methyl-1-propanol, 1-methylpropyl alcohol
PubChem CID 6568
ChEBI CHEBI:35687
IUPAC Name butan-2-ol
SMILES CCC(C)O

Specifications

Melting Point -115.0°C
Density 0.8100g/mL
Quantity 10 L
Flash Point 24°C
Infrared Spectrum Conforms to Ref Report
Packaging Plastic Drum
Physical Form Liquid
Refractive Index 1.3960 to 1.3980 (20°C, 589nm)
Residue after Evaporation 0.005% max.
Specific Gravity 0.81
Assay Percent Range 99%
Linear Formula C2H5CH(OH)CH3
Viscosity 4.2 mPa.s (20°C)
Water 0.5% max.
Beilstein 01, 371
Merck Index 15, 1543
Boiling Point 99.0°C
Solubility Information Solubility in water: 125g/L (20°C). Other solubilities: soluble in alcohol, ester, ether and benzene
Formula Weight 74.12
Percent Purity 99%
Grade Extra Pure
Chemical Name or Material sec-Butanol
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause drowsiness or dizziness.
Flammable liquid and vapor.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only