missing translation for 'onlineSavingsMsg'
Learn More

Agilent Technologies™ Seahorse XFp Glycolysis Stress Test Kit

Catalog No. 103017100
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
103017100 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 103017100 Supplier Agilent Technologies™ Supplier No. 103017100

Seahorse XFp Glycolysis Stress Test Kit containing six single-use pouches, each with glucose, oligomycin, and 2-deoxy-D-glucose. For use with Seahorse XF HS Mini and XFp analyzers.

  • Assay measures basal glycolysis and glycolysis capacity upon blockage of mitochondrial ATP production using respiratory modulators
  • Non-glycolytic acidification is assessed through glucose starvation
  • Compounds are reconstituted directly in the assay medium, eliminating the need for DMSO or other solvents
  • Each lot is rigorously tested and validated with functional assays to deliver consistent and robust results, eliminating the cost associated with failed experiments
  • Agilent™ Seahorse Report Generator simplifies data processing by automatically calculating the key parameters of your samples

Specifications

For Use With (Application) Cell Function
For Use With (Equipment) Other Fluorescence Detection Systems
Product Type Reagent Kit
Form Lyophilized
Quantity 1 Kit