missing translation for 'onlineSavingsMsg'
Learn More

Agilent Technologies™ Seahorse XFp Cell Culture Miniplate

Catalog No. 103025100
Change view
Click to view available options
No. of Wells:
8
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Wells
103025100 8
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 103025100 Supplier Agilent Technologies™ Supplier No. 103025100
Only null left

Seahorse XFp Cell Culture Miniplate is compatible with Seahorse XF HS Mini and XFp Analyzers and includes 12 Seahorse XFp Cell Culture Miniplates in individually sealed, gamma-irradiated tubs.

  • Available in a wide range of configurations to accomodate 2D monolayers, 3D spheroids, suspension cells, islets, and isolated mitochondria
  • Conforms to ANSI/SLAS Microplate Standards for footprint dimensions and well positions
  • Tissue culture treated for optimum adherence and growth of mammalian cells
  • Gamma-irradiated for use with live cell culture
  • Molded stops prevent the sensor cartridge sleeves from disrupting your cell monolayer
  • XF HS miniplate (XF HS Mini analyzer compatible) enables 1/3 fewer cells per well
  • Pre-coated Poly-D-Lysine cell culture plates, available in 96-well and 8-well formats, are available for use with XF assays using suspension cell, providing convenience and consistent assay result
  • PDL microplates are manufactured in an aseptic facility
  • Designed specifically for use with Seahorse XF Analyzers

Specifications

Barcode Includes Barcode
Format Standard
Array 1 x 8
Autoclavable Not Autoclavable
Color Clear
Disposable Single-use
For Use With (Equipment) Seahorse XF HS Mini Analyzer, Seahorse XFp Analyzer
Material Polystyrene
Volume (Metric) Well 250 μL
No. of Wells 8
Sterility Sterile
Surface Treatment Plasma
Well Shape Flat Bottom (F-well)
No. per Case 12 Assays/Cs.
Show More Show Less