missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SDS Solution, 20% Sodium Dodecyl Sulfate Solution, Molecular Biology/Electrophoresis, Fisher BioReagents™
$136.17 - $230.95
Chemical Identifiers
| CAS | 151-21-3 , 7732-18-5 |
|---|---|
| Molecular Formula | C12H25NaO4S |
| Molecular Weight (g/mol) | 288.38 |
| MDL Number | MFCD00036175 |
| InChI Key | DBMJMQXJHONAFJ-UHFFFAOYSA-M |
| Synonym | Sodium lauryl sulfate, SDS |
| PubChem CID | 3423265 |
| ChEBI | CHEBI:8984 |
| SMILES | [Na+].CCCCCCCCCCCCOS([O-])(=O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
BP1311200
|
Fisher BioReagents
V35202 |
200 mL | Poly Bottle |
Each for $136.17
Case of 6 Each for $817.02
|
|
||||
|
BP13111
|
Fisher BioReagents
BP1311 1 |
1 L | Nalgene Poly Bottle |
Each for $230.95
Case of 6 Each for $1,385.70
|
|
||||
Description
SDS Solution, 20% Sodium Dodecyl Sulfate Solution, Molecular Biology/Electrophoresis
- DNase, RNase, and protease-free premixed solution containing 20% SDS for protein extraction and DNA extraction procedures
- Reduce preparation time by using this premade SDS solution
- SDS is a commonly used detergent in protein purification, polyacrylamide gel electrophoresis (PAGE), and other molecular biology applications
- SDS is also used to disrupt cell membranes and dissociate nucleic acid-protein complexes in DNA extraction procedures
- Intended for research use only; not for human or animal diagnostic purposes
Chemical Identifiers
| C12H25NaO4S | |
| MFCD00036175 | |
| Sodium lauryl sulfate, SDS | |
| CHEBI:8984 |
Specifications
| 0.4 max. (0.1% solution) at 230nm, 0.1 max. (0.1% solution) at 280nm | |
| 151-21-3 , 7732-18-5 | |
| 100°C | |
| 19.5 to 20.5% | |
| (as Fatty Alcohol Sulfate): ≥99% | |
| CH3(CH2)10CH2OSO3Na | |
| 15, 8768 | |
| DBMJMQXJHONAFJ-UHFFFAOYSA-M | |
| sodium;dodecyl sulfate | |
| 3423265 | |
| 288.38 | |
| DNase-, RNase- and Protease-Free |
| Colorless | |
| Liquid | |
| 1.01g/cm³ | |
| Pass Test | |
| C12H25NaO4S | |
| MFCD00036175 | |
| Sodium lauryl sulfate, SDS | |
| [Na+].CCCCCCCCCCCCOS([O-])(=O)=O | |
| 288.38 | |
| CHEBI:8984 | |
| ≥99% | |
| Sodium Dodecyl Sulfate, 20% Solution |
Safety and Handling
Recommended Storage : RT