missing translation for 'onlineSavingsMsg'
Learn More

SDF4 Antibody, ProSci

Numéro de catalogue. 16822295
Modifier l'affichage
Click to view available options
Quantité:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
16822295 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 16822295 Fournisseur ProSci Llc Code fournisseur 424270100

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Goat Polyclonal Antibody

SDF4 Polyclonal Antibody specifically detects SDF4 in Human samples. It is validated for ELISA, Western Blot, Immunohistochemistry

Spécifications

Antigène SDF4
Applications ELISA, Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration 500 ug/mL
Conjugué Unconjugated
Dilution Peptide ELISA: antibody detection limit dilution 1:64000.Western Blot:Approx 45kDa band observed in lysates of cell line Jurkat (calculated MW of 41.8kDa according to NP_057260.2). Recommended concentration: 0.1-0.3ug/ml. Primary incubation was 1 hour. Preliminary testing was unsuccessful on Mouse for this particular batch.Immunohistochemistry:In paraffin embedded Human Kidney shows preferential staining in the brush borders of epithelial cells. Recommended concentration: 5-10ug/ml.
Formule Supplied at 0.5 mg/mL in Tris saline, 0.02% sodium azide, pH7.3 with 0.5% bovine serum albumin. Aliquot and store at -20°C. Minimize freezing and thawing.
Numéro d’acquisition du gène NP_057631.1,NP_057260.2
Alias du gène SDF4, stromal cell derived factor 4, Cab45, 45 kDa calcium-binding protein, OTTHUMP00000001754, SDF-4, calcium binding protein, stromal cell-derived factor 4
Espèces hôtes Goat
Méthode de purification Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
Quantité 0.1 mg
Statut réglementaire RUO
Primaire ou secondaire Primary
ID du gène (Entrez) 51150
Spécifications de test This antibody is expected to recognize both reported isoforms (NP_057631.1; NP_057260.2).
Espèces cibles Human
Contenu et stockage Aliquot and store at -20°C. Minimize freezing and thawing.
Type de produit Antibody
Forme Liquid
Isotype IgG
Catégorie de recherche Other
Afficher plus de résultats Afficher moins de résultats