Learn More
Agilent BioTek Scratch Assay App
GREENER_CHOICE
Catalog No. BT1750011
Change view
Click to view available options
For Use With (Equipment):
Scratch Assay Starter Kit (1750012)
Catalog No. For Use With (Equipment)
BT1750011 Scratch Assay Starter Kit (1750012)
1 options
Catalog No. BT1750011 Supplier Agilent BioTek Supplier No. 1750011
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Offers a simple interface for cell migration assay workflows, from kinetic capture through complete analysis. Pre-defined protocols for 24- and 96-well plates are available. Custom programming is quick and easy.
  • Offers a smooth, integrated workflow for kinetic image capture and powerful analysis of wound healing assays, performed with a Agilent BioTek Lionheart Automated Microscope or Cytation Cell Imaging Multi-Mode Reader
  • The Scratch Assay App and AutoScratch Wound Making Tool are critical components of the new Scratch Assay Starter Kit, which automates cell migration and wound healing assays
  • Simple installation and implementation
  • Pre-defined protocols for 24- and 96-well plate
  • Auto exposure and powerful image processing tools
  • Automatic statistics calculations: wound width, % confluence, maximum healing rate
  • For repeatable, consistent scratches and complete cell migration assay workflow consider the Scratch Assay Starter Kit
Specifications
For Use With (Application) Wound healing assays and Cell migration assays
Product Type Software
For Use With (Equipment) Scratch Assay Starter Kit (1750012)